Assay Details
| Gene Symbol: Rhox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022175.1 | NP_071511.1 | 3 | 453 | 117 | |
| XM_006257472.2 | XP_006257534.1 | 4 | 909 | 123 | ||
| XM_006257473.2 | XP_006257535.1 | 4 | 525 | 123 | ||
| XM_006257475.1 | XP_006257537.1 | 5 | 653 | 123 | ||
| XM_008773400.1 | XP_008771622.1 | 5 | 804 | 123 | ||
| GenBank mRNA | DQ058653.1 | - | 3 | 453 | 117 | |
| FQ228987.1 | - | 4 | 543 | 123 | ||
| S82627.1 | - | 3 | 445 | 123 | ||
More Information
Set Membership: |
|

Rhox5
Genomic Map