Assay Details
| Gene Symbol: Chrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031016.1 | NP_112278.1 | 1 | 2149 | 127 | |
| GenBank mRNA | J03025.1 | - | 1 | 2149 | 127 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of smooth muscle contraction
G-protein coupled acetylcholine receptor signaling pathway
chemical synaptic transmission
regulation of heart contraction
response to virus
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
drug binding
G-protein coupled acetylcholine receptor activity
regulation of smooth muscle contraction
G-protein coupled acetylcholine receptor signaling pathway
chemical synaptic transmission
regulation of heart contraction
response to virus
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
drug binding
G-protein coupled acetylcholine receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P267159 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P036514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76320 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P267159 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036514 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P267159 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036514 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P267159 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P036514 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P386689 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38922 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98227 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46275 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45742 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10674 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3757 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55173 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35259 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10854545 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854546 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854547 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10854544 | - | - |
| Rat Genome 230 2.0 Array | 1369245_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369245_at | - | - |
| Affymetrix HT Rat Focus Array | 1369245_at | - | - |
| Affymetrix HT Rat Focus Array | 1369245_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400004625.rn.1 | TC0400000642.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400004624.rn.1 | TC0400000642.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400004623.rn.1 | TC0400000642.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369245_PM_at | - | - |
| Rat Expression Set 230 | 1369245_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854544 | - | - |
| Rat Genome U34 Set | J03025_at | - | - |
| Rat Genome U34 Set | AB017655_s_at | - | - |
| Rat Genome U34 Set | AB017655_s_at | - | - |
| Rat Genome U34 Set | J03025_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21463923 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363635 | - | - |
| RatRef-12 Expression BeadChip | ILMN_54486 | - | - |

Chrm2
Genomic Map