Assay Details
| Gene Symbol: Tex21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025658.1 | NP_001020829.1 | 1 - 2 | 366 | 90 | |
| XM_008764744.1 | XP_008762966.1 | 2 - 3 | 484 | 90 | ||
| GenBank mRNA | BC097422.1 | - | 1 - 2 | 366 | 7454996 | 90 |
More Information
Updated Mapping Information
Set Membership: |
|

Tex21
Genomic Map