Assay Details
| Gene Symbol: Fam221a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025771.2 | NP_001020942.1 | 5 - 6 | 762 | 63 | |
| XM_006236467.2 | XP_006236529.1 | 5 - 6 | 735 | 63 | ||
| XM_006236468.2 | XP_006236530.1 | 4 - 5 | 749 | 63 | ||
| GenBank mRNA | BC097434.1 | - | 5 - 6 | 763 | 7455430 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Fam221a
Genomic Map