Assay Details
| Gene Symbol: Zdhhc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039014.1 | NP_001034103.1 | 3 - 4 | 551 | 66 | |
| XM_006244188.2 | XP_006244250.1 | 4 - 5 | 724 | 66 | ||
| XM_008766714.1 | XP_008764936.1 | 4 - 5 | 728 | 66 | ||
| XM_008766715.1 | XP_008764937.1 | 4 - 5 | 1113 | 66 | ||
| XM_008766716.1 | XP_008764938.1 | 5 - 6 | 939 | 66 | ||
| GenBank mRNA | AY886522.1 | - | 3 - 4 | 551 | 66 | |
| BC128697.1 | - | 4 - 5 | 670 | 7444822 | 66 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zdhhc3
Genomic Map