Assay Details
| Gene Symbol: Pced1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039454.1 | NP_001034543.1 | 5 | 1543 | 60 | |
| XM_008765737.1 | XP_008763959.1 | 2 | 1835 | 60 | ||
| XM_008765738.1 | XP_008763960.1 | 2 | 1832 | 60 | ||
| XM_008765739.1 | XP_008763961.1 | 2 | 1729 | 60 | ||
| XM_008765740.1 | XP_008763962.1 | 1 | 1194 | 60 | ||
| XM_008765741.1 | XP_008763963.1 | 3 | 2105 | 60 | ||
| XM_008765742.1 | XP_008763964.1 | 2 | 2482 | 60 | ||
| XM_008765743.1 | XP_008763965.1 | 2 | 1726 | 60 | ||
| XM_008765744.1 | XP_008763966.1 | 2 | 2479 | 60 | ||
| GenBank mRNA | BC112322.1 | - | 5 | 1543 | 7936449 | 60 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P043589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166175 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P043589 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P043589 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P043589 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P131230 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P996952 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82608 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56875 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15959 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42921 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5378 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38245 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76286 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42138 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10898944 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898945 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898946 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898941 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898942 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10898940 | - | - |
| Rat Genome 230 2.0 Array | 1374355_at | - | - |
| Rat Genome 230 2.0 Array | 1374355_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374355_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374355_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700013436.rn.1 | TC0700001747.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700013438.rn.1 | TC0700001747.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700013452.rn.1 | TC0700001747.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700013450.rn.1 | TC0700001747.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700013451.rn.1 | TC0700001747.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700013444.rn.1 | TC0700001747.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700013440.rn.1 | TC0700001747.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700013449.rn.1 | TC0700001747.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374355_at | - | - |
| Rat Expression Set 230 | 1374355_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898940 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370472 | - | - |

Pced1b
Genomic Map