Assay Details
| Gene Symbol: RGD1359290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047898.1 | NP_001041363.1 | 3 - 4 | 437 | 126 | |
| XM_008768380.1 | XP_008766602.1 | 1 - 2 | 542 | 126 | ||
| GenBank mRNA | AY321338.1 | - | 3 - 4 | 437 | 126 | |
More Information
Set Membership: |
|

RGD1359290
Genomic Map