Assay Details
| Gene Symbol: LOC500846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047962.1 | NP_001041427.1 | 6 - 7 | 1020 | 57 | |
| GenBank mRNA | AY539897.1 | - | 6 - 7 | 1020 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC500846
Genomic Map