Assay Details
| Gene Symbol: LOC501618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047970.1 | NP_001041435.1 | 1 - 2 | 79 | 109 | |
| GenBank mRNA | AY539930.1 | - | 1 - 2 | 79 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC501618
Genomic Map