Assay Details
| Gene Symbol: Ccdc184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024909.1 | NP_001020080.1 | 1 | 843 | 79 | |
| GenBank mRNA | BC097418.1 | - | 1 | 843 | 7454547 | 79 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccdc184
Genomic Map