Assay Details
| Gene Symbol: LOC367195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047920.1 | NP_001041385.1 | 2 | 169 | 140 | |
| GenBank mRNA | AY325186.1 | - | 2 | 169 | 140 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC367195
Genomic Map