Assay Details
| Gene Symbol: MT-CYB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | NC_001665.CYTB.0 | - | 953 | 77 | ||
More Information
Additional Information:
NC_001665 (strand:+; 14136..15278); mitochondrially encoded cytochrome b
Set Membership: |
|

MT-CYB
Genomic Map