Assay Details
| Gene Symbol: Tmem150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107476.1 | NP_001100946.1 | 4 - 5 | 625 | 67 | |
| XM_006228267.2 | XP_006228329.1 | 5 - 6 | 1035 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem150b
Genomic Map