Assay Details
| Gene Symbol: Lrrc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135896.1 | NP_001129368.1 | 3 - 4 | 436 | 97 | |
| XM_008765581.1 | XP_008763803.1 | 1 - 2 | 254 | 97 | ||
| XM_008765582.1 | XP_008763804.1 | 1 - 2 | 273 | 97 | ||
| XM_008765583.1 | XP_008763805.1 | 2 - 3 | 977 | 97 | ||
| GenBank mRNA | BC127521.1 | - | 3 - 4 | 436 | 7110443 | 97 |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc24
Genomic Map