Assay Details
| Gene Symbol: Lrtomt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139494.1 | NP_001132966.1 | 1 - 2 | 276 | 64 | |
| GenBank mRNA | EU627093.1 | - | 1 - 2 | 276 | 64 | |
More Information
Set Membership: |
|

Lrtomt
Genomic Map