Assay Details
| Gene Symbol: Zbtb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134991.1 | NP_001128463.1 | 2 | 849 | 99 | |
| GenBank mRNA | BC169087.1 | - | 2 | 849 | 5600062 | 99 |
More Information
Set Membership: |
|

Zbtb12
Genomic Map