Assay Details
| Gene Symbol: Nlrp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169149.1 | NP_001162620.1 | 4 - 5 | 2176 | 89 | |
| XM_006228188.1 | XP_006228250.1 | 3 - 4 | 2011 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nlrp9
Genomic Map