Assay Details
| Gene Symbol: Cidea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170467.1 | NP_001163938.1 | 4 - 5 | 512 | 67 | |
| GenBank mRNA | BF284899.1 | - | 67 | |||
| FM041444.1 | - | 4 - 5 | 512 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cidea
Genomic Map