Assay Details
| Gene Symbol: Smim14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037792.1 | NP_001032881.1 | 1 - 2 | 86 | 108 | |
| GenBank mRNA | BC100271.1 | - | 1 - 2 | 86 | 7445571 | 108 |
| FQ229716.1 | - | 1 - 2 | 145 | 108 | ||
| FQ229969.1 | - | 1 - 2 | 97 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC100271.1, NM_001037792.1 transcripts.
Set Membership: |
|

Smim14
Genomic Map