Assay Details
| Gene Symbol: Smim14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037792.1 | NP_001032881.1 | 2 - 3 | 200 | 76 | |
| XM_006250961.2 | XP_006251023.1 | 2 - 3 | 458 | 76 | ||
| GenBank mRNA | BC100271.1 | - | 2 - 3 | 200 | 7445571 | 76 |
| FQ229716.1 | - | 2 - 3 | 259 | 76 | ||
| FQ229847.1 | - | 1 - 2 | 84 | 76 | ||
| FQ229969.1 | - | 2 - 3 | 211 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Smim14
Genomic Map