Assay Details
| Gene Symbol: Mir27b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031832.1 | 1 | 49 | 69 | ||
| Gene Symbol: Npepo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771429.1 | XP_008769651.1 | 17 | 4846 | 69 | |
| XM_008771431.1 | XP_008769653.1 | 16 | 4693 | 69 | ||
| XM_008771432.1 | XP_008769654.1 | 16 | 4685 | 69 | ||
| XM_008771433.1 | XP_008769655.1 | 16 | 4558 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 23b | Mir23b | rno-mir-23b | -143.0 | Sense | Chr. 17 - 823222 - 823318 on NCBI Build Rnor_6.0 |
| Downstream | microRNA 3074 | Mir3074 | rno-mir-3074 | 383.0 | Anti-sense | Chr. 17 - 823940 - 824056 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of gene expression
Wnt signaling pathway
myelination
positive regulation of angiogenesis
retina vasculature development in camera-type eye
cellular response to inorganic substance
regulation of semaphorin-plexin signaling pathway
proteolysis
leukotriene biosynthetic process
aminopeptidase activity
metallopeptidase activity
zinc ion binding
regulation of gene expression
Wnt signaling pathway
myelination
positive regulation of angiogenesis
retina vasculature development in camera-type eye
cellular response to inorganic substance
regulation of semaphorin-plexin signaling pathway
proteolysis
leukotriene biosynthetic process
aminopeptidase activity
metallopeptidase activity
zinc ion binding
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P231867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136579 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P231867 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P231867 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P231867 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P431354 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P231867 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11056 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85546 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23309 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50439 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22957 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103239 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36874 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70892 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30941 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98731 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3274 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3568 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 95996 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67073 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31287 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27300 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53658 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80788 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36939 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91912 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45436 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92835 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10796993 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797004 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796995 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796997 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797000 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797003 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796998 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797005 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796996 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796994 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797001 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797006 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796992 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796999 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797002 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10796991 | - | - |
| Rat Genome 230 2.0 Array | 1374767_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374767_at | - | - |
| Affymetrix HT Rat Focus Array | 1374767_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374767_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700000100.rn.1 | TC1700000015.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374767_at | - | - |
| Rat Expression Set 230 | 1374767_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796991 | - | - |
| Rat Genome U34 Set | rc_AA799396_g_at | - | - |
| Rat Genome U34 Set | rc_AA799396_at | - | - |
| Rat Genome U34 Set | AA799397_at | - | - |
| Rat Genome U34 Set | rc_AI072900_at | - | - |

Mir27b
Genomic Map