Assay Details
| Gene Symbol: Mir196b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031946.1 | 1 | 44 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeobox protein Hox-A9 | LOC103689925 | -3.9kb | Sense | Chr. 4 - 82191804 - 82195162 on NCBI Build Rnor_6.0 | |
| Downstream | homeobox protein Hox-A10-like | LOC100912608 | 1.0kb | Sense | Chr. 4 - 82200159 - 82210294 on NCBI Build Rnor_6.0 |
More Information
Set Membership: |
|

Mir196b
Genomic Map