Assay Details
| Gene Symbol: Mir10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031815.1 | 1 | 54 | 89 | ||
| Gene Symbol: Hoxd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761882.1 | XP_008760104.1 | 2 | 4932 | 89 | |
| XM_008761883.1 | XP_008760105.1 | 2 | 3659 | 89 | ||
| XM_008761884.1 | XP_008760106.1 | 3 | 4411 | 89 | ||
| XM_008761885.1 | XP_008760107.1 | 3 | 4833 | 89 | ||
| XM_008761886.1 | XP_008760108.1 | 3 | 2846 | 89 | ||
| XM_008761887.1 | XP_008760109.1 | 4 | 2747 | 89 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeobox protein Hox-D8-like | LOC100912936 | -18.2kb | Sense | Chr. 3 - 61627329 - 61629743 on NCBI Build Rnor_6.0 | |
| Downstream | homeo box D3 | Hoxd3 | Hox4r6 | 2.5kb | Sense | Chr. 3 - 61650588 - 61670285 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008761882.1, XM_008761883.1, XM_008761884.1, XM_008761885.1, XM_008761886.1, XM_008761887.1 transcripts.
Set Membership: |
|

Mir10b
Genomic Map