Assay Details
| Gene Symbol: Cldn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107135.2 | NP_001100605.1 | 3 - 4 | 463 | 82 | |
| XM_006249155.2 | XP_006249217.1 | 4 - 5 | 2111 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cldn15
Genomic Map