Assay Details
| Gene Symbol: RGD1311756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173557.1 | NP_001167028.1 | 11 - 12 | 1755 | 64 | |
| XM_006240439.2 | XP_006240501.1 | 12 - 13 | 1929 | 64 | ||
| XM_006240441.2 | XP_006240503.1 | 12 - 13 | 1990 | 64 | ||
| XM_006240442.2 | XP_006240504.1 | 10 - 11 | 2742 | 64 | ||
| XM_008764817.1 | XP_008763039.1 | 12 - 13 | 1984 | 64 | ||
| GenBank mRNA | FQ221290.1 | - | 10 - 11 | 1611 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1311756
Genomic Map