Assay Details
| Gene Symbol: RGD1311756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173557.1 | NP_001167028.1 | 12 - 13 | 1870 | 58 | |
| XM_006240439.2 | XP_006240501.1 | 13 - 14 | 2044 | 58 | ||
| XM_006240441.2 | XP_006240503.1 | 13 - 14 | 2105 | 58 | ||
| XM_006240442.2 | XP_006240504.1 | 11 - 12 | 2857 | 58 | ||
| XM_008764817.1 | XP_008763039.1 | 13 - 14 | 2099 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RGD1311756
Genomic Map