Assay Details
| Gene Symbol: RGD1311756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173557.1 | NP_001167028.1 | 5 - 6 | 775 | 64 | |
| XM_006240439.2 | XP_006240501.1 | 6 - 7 | 949 | 64 | ||
| XM_006240441.2 | XP_006240503.1 | 6 - 7 | 1010 | 64 | ||
| XM_006240442.2 | XP_006240504.1 | 4 - 5 | 1762 | 64 | ||
| XM_008764817.1 | XP_008763039.1 | 6 - 7 | 1004 | 64 | ||
| GenBank mRNA | FQ221290.1 | - | 4 - 5 | 631 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1311756
Genomic Map