Assay Details
| Gene Symbol: RGD1311756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173557.1 | NP_001167028.1 | 6 - 7 | 919 | 90 | |
| XM_006240439.2 | XP_006240501.1 | 7 - 8 | 1093 | 90 | ||
| XM_006240441.2 | XP_006240503.1 | 7 - 8 | 1154 | 90 | ||
| XM_006240442.2 | XP_006240504.1 | 5 - 6 | 1906 | 90 | ||
| XM_008764817.1 | XP_008763039.1 | 7 - 8 | 1148 | 90 | ||
| GenBank mRNA | FQ221290.1 | - | 5 - 6 | 775 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1311756
Genomic Map