Assay Details
| Gene Symbol: RGD1311756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173557.1 | NP_001167028.1 | 8 - 9 | 1241 | 62 | |
| XM_006240439.2 | XP_006240501.1 | 9 - 10 | 1415 | 62 | ||
| XM_006240441.2 | XP_006240503.1 | 9 - 10 | 1476 | 62 | ||
| XM_006240442.2 | XP_006240504.1 | 7 - 8 | 2228 | 62 | ||
| XM_008764817.1 | XP_008763039.1 | 9 - 10 | 1470 | 62 | ||
| GenBank mRNA | FQ221290.1 | - | 7 - 8 | 1097 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1311756
Genomic Map