Assay Details
| Gene Symbol: Sp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191774.1 | NP_001178703.1 | 2 | 218 | 65 | |
| XM_008765001.1 | XP_008763223.1 | 2 | 304 | 65 | ||
| XM_008765002.1 | XP_008763224.1 | 2 | 227 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sp8
Genomic Map