Assay Details
| Gene Symbol: Fbxo28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107203.3 | NP_001100673.1 | 1 - 2 | 281 | 74 | |
| GenBank mRNA | BC167003.1 | - | 1 - 2 | 282 | 8366732 | 74 |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo28
Genomic Map