Assay Details
| Gene Symbol: Fbxo28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107203.3 | NP_001100673.1 | 3 - 4 | 533 | 93 | |
| GenBank mRNA | BC167003.1 | - | 3 - 4 | 534 | 8366732 | 93 |
| FQ223120.1 | - | 2 - 3 | 229 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo28
Genomic Map