Assay Details
| Gene Symbol: Rasd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270954.1 | NP_001257883.1 | 1 - 2 | 375 | 61 | |
| XM_008767871.1 | XP_008766093.1 | 2 - 3 | 2327 | 61 | ||
| XM_008767872.1 | XP_008766094.1 | 2 - 3 | 2331 | 61 | ||
| XM_008767873.1 | XP_008766095.1 | 3 - 4 | 4980 | 61 | ||
| GenBank mRNA | AF239157.1 | - | 1 - 2 | 403 | 61 | |
| BC099136.1 | - | 1 - 2 | 375 | 7455971 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rasd1
Genomic Map