Assay Details
| Gene Symbol: Egr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252240.2 | XP_006252302.1 | 2 - 3 | 480 | 77 | |
| XM_008770818.1 | XP_008769040.1 | 1 - 2 | 240 | 77 | ||
| GenBank mRNA | BC097334.1 | - | 448 | 7456458 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC097334.1, XM_006252240.2, XM_008770818.1 transcripts.
Set Membership: |
|

Egr3
Genomic Map