Assay Details
Gene Symbol: LOC498316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109085.1 | NP_001102555.1 | 2 | 178 | 66 | |
XM_006250489.2 | XP_006250551.1 | 2 | 238 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
