Assay Details
| Gene Symbol: Csrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177425.3 | NP_803174.2 | 4 - 5 | 464 | 88 | |
| GenBank mRNA | BC086356.1 | - | 4 - 5 | 478 | 7303714 | 88 |
| U44948.1 | - | 4 - 5 | 469 | 88 | ||
More Information
Set Membership: |
|

Csrp2
Genomic Map