Assay Details
| Gene Symbol: Tmub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235944.2 | XP_006236006.1 | 1 | 114 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006235944.2 transcripts.
Set Membership: |
|

Tmub1
Genomic Map