Assay Details
| Gene Symbol: Kbtbd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749799.3 | XP_003749847.1 | 1 | 90 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003749799.3 transcripts.
Set Membership: |
|

Kbtbd12
Genomic Map