Assay Details
| Gene Symbol: Fam159a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001070397.5 | XP_001070397.2 | 2 - 3 | 491 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam159a| Gene Symbol: Fam159a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001070397.5 | XP_001070397.2 | 2 - 3 | 491 | 58 | |
Set Membership: |
|