Assay Details
| Gene Symbol: Abhd17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014028.1 | NP_001014050.1 | 4 | 1395 | 121 | |
| XM_006231162.2 | XP_006231224.1 | 4 | 1485 | 121 | ||
| GenBank mRNA | BC079229.1 | - | 4 | 1395 | 7132456 | 121 |
| FQ227098.1 | - | 3 | 743 | 121 | ||
More Information
Set Membership: |
|

Abhd17b
Genomic Map