Assay Details
| Gene Symbol: Mafg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247912.2 | XP_006247974.1 | 3 | 7672 | 95 | |
| XM_006247913.2 | XP_006247975.1 | 4 | 6673 | 95 | ||
| XM_006247914.2 | XP_006247976.1 | 2 | 3592 | 95 | ||
| XM_006247916.2 | XP_006247978.1 | 2 | 5369 | 95 | ||
| XM_006247917.2 | XP_006247979.1 | 2 | 8141 | 95 | ||
| XM_006247918.2 | XP_006247980.1 | 2 | 4752 | 95 | ||
| XM_006247919.2 | XP_006247981.1 | 2 | 5299 | 95 | ||
| XM_006247921.2 | XP_006247983.1 | 3 | 4350 | 95 | ||
| XM_006247922.2 | XP_006247984.1 | 3 | 3511 | 95 | ||
| XM_006247923.2 | XP_006247985.1 | 3 | 4280 | 95 | ||
| XM_008768482.1 | XP_008766704.1 | 3 | 7122 | 95 | ||
More Information
Set Membership: |
|

Mafg
Genomic Map