Assay Details
| Gene Symbol: Map3k10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191621.3 | NP_001178550.1 | 10 | 3231 | 106 | |
| XM_006228576.2 | XP_006228638.1 | 10 | 3434 | 106 | ||
More Information
Set Membership: |
|

Map3k10
Genomic Map