Assay Details
| Gene Symbol: LOC102554944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248802.2 | XP_006248864.1 | 2 - 3 | 262 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102554944| Gene Symbol: LOC102554944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248802.2 | XP_006248864.1 | 2 - 3 | 262 | 92 | |
Set Membership: |
|