Assay Details
| Gene Symbol: Upk3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249208.2 | XP_006249270.1 | 3 - 4 | 642 | 84 | |
| XM_341056.7 | XP_341057.3 | 3 - 4 | 642 | 84 | ||
More Information
Set Membership: |
|

Upk3b
Genomic Map