Assay Details
| Gene Symbol: Upk3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249208.2 | XP_006249270.1 | 4 - 5 | 720 | 78 | |
| XM_341056.7 | XP_341057.3 | 4 - 5 | 720 | 78 | ||
More Information
Set Membership: |
|

Upk3b
Genomic Map