Assay Details
| Gene Symbol: Tmem59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139465.2 | NP_001132937.1 | 8 | 1084 | 109 | |
| GenBank mRNA | BF287725.1 | - | 4 | 433 | 109 | |
| CV126271.1 | - | 2 | 261 | 109 | ||
| FM091711.1 | - | 4 | 504 | 109 | ||
| FM102590.1 | - | 1 | 88 | 109 | ||
| FQ212504.1 | - | 8 | 1050 | 109 | ||
| FQ214069.1 | - | 8 | 1082 | 109 | ||
| FQ226828.1 | - | 7 | 804 | 109 | ||
| FQ231829.1 | - | 8 | 1070 | 109 | ||
| FQ232106.1 | - | 8 | 1089 | 109 | ||
More Information
Set Membership: |
|

Tmem59
Genomic Map