Assay Details
| Gene Symbol: Med18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115037.1 | NP_001108509.1 | 3 | 1211 | 105 | |
| XM_008764174.1 | XP_008762396.1 | 2 | 1053 | 105 | ||
| GenBank mRNA | BC158672.1 | - | 3 | 1124 | 8371540 | 105 |
More Information
Set Membership: |
|

Med18
Genomic Map