Assay Details
| Gene Symbol: RGD1564664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110055.1 | NP_001103525.1 | 1 | 624 | 77 | |
| GenBank mRNA | AB010079.1 | - | 1 | 624 | 77 | |
More Information
Set Membership: |
|

RGD1564664
Genomic Map