Assay Details
| Gene Symbol: Fignl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011913.1 | NP_001011913.1 | 2 - 3 | 160 | 83 | |
| XM_006251477.2 | XP_006251539.1 | 2 - 3 | 393 | 83 | ||
| XM_008770261.1 | XP_008768483.1 | 2 - 3 | 207 | 83 | ||
| GenBank mRNA | AY623031.1 | - | 2 - 3 | 160 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY623031.1, NM_001011913.1, XM_006251477.2, XM_008770261.1 transcripts.
Set Membership: |
|

Fignl1
Genomic Map