Assay Details
| Gene Symbol: Zcchc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013156.1 | NP_001013174.1 | 3 - 4 | 660 | 60 | |
| XM_006231841.2 | XP_006231903.1 | 4 - 5 | 762 | 60 | ||
| GenBank mRNA | BC083695.1 | - | 3 - 4 | 660 | 7189545 | 60 |
More Information
Set Membership: |
|

Zcchc9
Genomic Map